Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337854

Sigma-Aldrich

4-Chloro-2,6-dinitroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(NO2)2NH2
CAS Number:
Molecular Weight:
217.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

145-147 °C (lit.)

SMILES string

Nc1c(cc(Cl)cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4ClN3O4/c7-3-1-4(9(11)12)6(8)5(2-3)10(13)14/h1-2H,8H2

InChI key

CLMQUEQFVUMDPC-UHFFFAOYSA-N

General description

Standard molar enthalpy of formation of 4-chloro-2,6-dinitroaniline has been evaluated.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental thermochemical study of two chlorodinitroaniline isomers.
Ribeiro da Silva MAV, et al.
The Journal of Chemical Thermodynamics, 42(4), 496-501 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service