Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337633

Sigma-Aldrich

4-Chloro-2,8-bis(trifluoromethyl)quinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H4ClF6N
CAS Number:
Molecular Weight:
299.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

46-48 °C (lit.)

functional group

chloro
fluoro

SMILES string

FC(F)(F)c1cc(Cl)c2cccc(c2n1)C(F)(F)F

InChI

1S/C11H4ClF6N/c12-7-4-8(11(16,17)18)19-9-5(7)2-1-3-6(9)10(13,14)15/h1-4H

InChI key

ZSQOESPYYNJBCZ-UHFFFAOYSA-N

General description

4-Chloro-2,8-bis(trifluoromethyl)quinolone is the intermediate formed during the synthesis of quinoline derivatives.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumesh Eswaran et al.
European journal of medicinal chemistry, 45(8), 3374-3383 (2010-06-12)
Four new series of quinoline derivatives were synthesized starting from 2-trifluoromethyl aniline through multi-step reactions. In the reaction sequence, substituted aniline was cyclized to 4-hydroxy quinoline 1, which was then transformed to 4-chloro-2,8-bis(trifluoromethyl)quinoline 2. The key scaffold 4-hydrazinyl-2,8-bis(trifluoromethyl)quinoline 3, obtained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service