Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337609

Sigma-Aldrich

Glycocholic acid-(glycyl-1-13C) monohydrate

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC25H43NO6 · H2O
CAS Number:
Molecular Weight:
484.63
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

135-140 °C (lit.)

mass shift

M+1

SMILES string

O.C[C@H](CCC(=O)NC[13C](O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C26H43NO6.H2O/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29;/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33);1H2/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-;/m1./s1/i23+1;

InChI key

WDKPRHOCWKLQPK-TXDHSPGBSA-N

Related Categories

Application

Substrate used for 13CO2 breath tests in ileal dysfunction diagnosis.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G W Hepner
Gastroenterology, 68(1), 8-16 (1975-01-01)
Deconjugation of bile salts was assessed by measuring the rate of appearance of 14-CO2 in the breath after the administration of cholyl [1-14-C]glycine to patients with ileal dysfunction (ileitis or ileal resection), cholesterol cholelithiasis with or without cholecystectomy, and control
Ximena Mora-Cubillos et al.
Molecular nutrition & food research, 59(12), 2480-2490 (2015-09-29)
To identify the most discriminant dietary biomarkers of nuts exposure in subjects with metabolic syndrome (MetS), and investigate the potential association between exposure and the severity of the MetS diagnostic traits. We applied the untargeted LC-ESI-qToF-MS-driven metabolomic workflow to explore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service