Skip to Content
MilliporeSigma
All Photos(1)

Documents

337110

Sigma-Aldrich

2-Fluoro-4-methylaniline

99%

Synonym(s):

2-Fluoro-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)NH2
CAS Number:
Molecular Weight:
125.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

70-71 °C/7 mmHg (lit.)

density

1.108 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)c(F)c1

InChI

1S/C7H8FN/c1-5-2-3-7(9)6(8)4-5/h2-4H,9H2,1H3

InChI key

ZQEXBVHABAJPHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rat liver microsomal metabolism of 2-fluoro-4-methylaniline was studied by HPLC.

Application

2-Fluoro-4-methylaniline was used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It was also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Boeren et al.
Xenobiotica; the fate of foreign compounds in biological systems, 22(12), 1403-1423 (1992-12-01)
1. Rat liver microsomal metabolism of 2-fluoro-, 2-chloro- and 2-bromo-4-methylaniline was investigated using h.p.l.c. Metabolites identified include products from side-chain C-hydroxylation (benzyl alcohols and benzaldehydes) and N-hydroxylation (hydroxylamines and nitroso derivatives). Aromatic ring hydroxylation was not a major reaction pathway.
Sharon T Marino et al.
Molecules (Basel, Switzerland), 9(6), 405-426 (2007-11-17)
In the past decade there has been a significant growth in the sales of pharmaceutical drugs worldwide, but more importantly there has been a dramatic growth in the sales of single enantiomer drugs. The pharmaceutical industry has a rising demand
J M Bentley et al.
Bioorganic & medicinal chemistry letters, 14(9), 2367-2370 (2004-04-15)
A series of 1-(1-indolinyl)-2-propylamines was synthesised and evaluated as 5-HT(2C) receptor agonists for the treatment of obesity. The general methods of synthesis of the precursor indoles are described. The functional efficacy and radioligand binding data for all of the compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service