Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

336491

Sigma-Aldrich

O-(4-Chlorobenzoyl)hydroquinine

98%

Synonym(s):

Dihydroquinine 4-chlorobenzoate, Hydroquinine 4-chlorobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29ClN2O3
CAS Number:
Molecular Weight:
464.98
Beilstein:
4302318
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]26/D +150°, c = 1 in ethanol

mp

128-130 °C (lit.)

functional group

chloro
ester

SMILES string

CC[C@H]1CN2CCC1CC2[C@H](OC(=O)c3ccc(Cl)cc3)c4ccnc5ccc(OC)cc45

InChI

1S/C27H29ClN2O3/c1-3-17-16-30-13-11-19(17)14-25(30)26(33-27(31)18-4-6-20(28)7-5-18)22-10-12-29-24-9-8-21(32-2)15-23(22)24/h4-10,12,15,17,19,25-26H,3,11,13-14,16H2,1-2H3/t17-,19-,25-,26+/m0/s1

InChI key

TXVNNFDXQZFMBQ-MFQNCIFPSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Sold under license from Rhodia Pharma Solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service