Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

336386

Sigma-Aldrich

trans-3-Furanacrylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

155-158 °C (lit.)

SMILES string

OC(=O)\C=C\c1ccoc1

InChI

1S/C7H6O3/c8-7(9)2-1-6-3-4-10-5-6/h1-5H,(H,8,9)/b2-1+

InChI key

JHAPZUDWRRBZHZ-OWOJBTEDSA-N

Application

trans-3-Furanacrylic acid was used in the detection of cis-urocanic acid in serum of mice by immunoassay.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Moodycliffe et al.
Immunology, 79(4), 667-672 (1993-08-01)
Cis-urocanic acid (cis-UCA), which is formed from the naturally occurring trans-isomer on ultraviolet (UV) irradiation, has been suggested as a photoreceptor for and mediator of the suppressive effects of UV irradiation on systemic immune responses. Trans-UCA is located predominantly in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service