Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

336017

Sigma-Aldrich

4-Aminophthalonitrile

98%

Synonym(s):

3,4-Dicyanoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-1,2-(CN)2
CAS Number:
Molecular Weight:
143.15
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

98%

form

powder, crystals or chunks

mp

179-181 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc(C#N)c(c1)C#N

InChI

1S/C8H5N3/c9-4-6-1-2-8(11)3-7(6)5-10/h1-3H,11H2

InChI key

RRCAJFYQXKPXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service