Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335800

Sigma-Aldrich

Ethyl 2-bromobenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CO2C2H5
CAS Number:
Molecular Weight:
229.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.544 (lit.)

bp

117 °C/17 mmHg (lit.)

density

1.443 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CCOC(=O)c1ccccc1Br

InChI

1S/C9H9BrO2/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6H,2H2,1H3

InChI key

BIHHBTVQFPVSTE-UHFFFAOYSA-N

General description

Ethyl 2-bromobenzoate is an halobenzoic acid ethyl ester. Silylation and hydrolysis of ethyl 2-bromobenzoate has been reported.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gernot A Eller
Molecules (Basel, Switzerland), 11(11), 915-928 (2007-11-17)
This work deals with the use of organic systematic nomenclature in scientific literature, its quality, and computerized methods for its improvement. Criteria for classification of systematic names in terms of quality/correctness are discussed and applied to a sample set of
Scott E Denmark et al.
Organic letters, 5(19), 3483-3486 (2003-09-12)
[reaction: see text] A mild and general palladium-catalyzed insertion of 1,2-diethoxy-1,1,2,2-tetramethyldisilane into a variety of aryl bromides affords the aryldimethylsilyl ethers in high yields. Hydrolysis of the ethers under pH-optimized conditions results in the exclusive formation of the desired aryldimethylsilanols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service