Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335762

Sigma-Aldrich

1-Bromo-4-butylbenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H4Br
CAS Number:
Molecular Weight:
213.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.53 (lit.)

density

1.208 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CCCCc1ccc(Br)cc1

InChI

1S/C10H13Br/c1-2-3-4-9-5-7-10(11)8-6-9/h5-8H,2-4H2,1H3

InChI key

BRGVKVZXDWGJBX-UHFFFAOYSA-N

General description

Phosphite or phosphine oxide ligand catalyzed Suzuki-Miyaura reaction of 2-pyridyl boron derivatives with 1-bromo-4-butylbenzene has been reported.[1]

Application

1-Bromo-4-butylbenzene was used in the preparation of:
  • 4-(4-butylphenyl)-2-methylbut-3-yn-2-ol[2]
  • teraaza- and hexaazacyclophane[3]
  • 4-butyltriphenylamine, monomer required for the preparation of poly(4-butyltriphenylamine)[4]

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A general and efficient method for the Suzuki-Miyaura coupling of 2-pyridyl nucleophiles.
Kelvin L Billingsley et al.
Angewandte Chemie (International ed. in English), 47(25), 4695-4698 (2008-05-21)
Irena Kulszewicz-Bajer et al.
Physical chemistry chemical physics : PCCP, 11(9), 1362-1368 (2009-02-19)
Teraaza- and hexaazacyclophanes have been synthesized via a one-step palladium catalyzed amination reaction. The chemical oxidation of cyclophanes leads to the formation of radical cations, the presence of which was manifested by the appearance of new bands in UV-Vis-NIR and
Preparation of poly (4-butyltriphenylamine) particles by chemical oxidative dispersion polymerization.
Fujioka M, et al.
Coll. Polymer Sci., 286(3), 313-318 (2008)
Synthesis and characterization of the multi-photon absorption and excited-state properties of a neat liquid 4-propyl 4'-butyl diphenyl acetylene
Khoo LC, et al.
Journal of Materials Chemistry, 19(40), 7525-7525 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service