Skip to Content
MilliporeSigma
All Photos(1)

Documents

335487

Sigma-Aldrich

5′-Nitro-2′-(4-nitrophenyl)benzanilide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHC6H3(NO2)C6H4NO2
CAS Number:
Molecular Weight:
363.32
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

98%

mp

239-242 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(cc1)-c2ccc(cc2NC(=O)c3ccccc3)[N+]([O-])=O

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service