Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335274

Sigma-Aldrich

2,2-Dimethyl-3(2H)-furanone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

liquid

refractive index

n20/D 1.455 (lit.)

bp

46-47 °C/18 mmHg (lit.)

density

1.027 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)OC=CC1=O

InChI

1S/C6H8O2/c1-6(2)5(7)3-4-8-6/h3-4H,1-2H3

InChI key

HVNICCSXGONRCB-UHFFFAOYSA-N

General description

2,2-Dimethyl-3(2H)-furanone is an β-oxo enol ether.[1] Conjugate addition of 2,2-dimethyl-3(2H)-furanone to organocuprates has been reported.[2] Photoannelation of alkenes to 2,2-dimethyl-3(2H)-furanone yields cyclohexenone.[3]

Application

2,2-Dimethyl-3(2H)-furanone was used in the synthesis of epoxides via epoxidation.[1] It was also used in the preparation of racemic methyl nonactate.[4]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethyldioxirane epoxidation of ?-oxo enol ethers.
Adam W and Hadjiarapoglou L.
Chemische Berichte, 123(10), 2077-2079 (1990)
Stereoselective synthesis of (.+-.) methyl nonactate.
Baldwin SW and McIver JM.
The Journal of Organic Chemistry, 52(2), 320-322 (1987)
Conjugated Addition to 2, 2-Dimethyl-3 (2h)-furanone: Support for the Baldwin Rules for Ring Closure.
Smith AB and Jerris PJ.
Synthetic Communications, 8(7), 421-426 (1978)
Cyclohexenones by the photoannelation of alkenes with 2, 2-dimethyl-3 (2H)-furanone.
Baldwin SW and Wilkinson JM.
Tetrahedron Letters, 20(29), 2657-2660 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service