334847
N-Fluoro-N-propyl-p-toluenesulfonamide
technical grade, 80%
About This Item
grade
technical grade
Assay
80%
SMILES string
CCCN(F)S(=O)(=O)c1ccc(C)cc1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service