Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

334669

Sigma-Aldrich

2-Amino-5-(ethylthio)-1,3,4-thiadiazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3S2
CAS Number:
Molecular Weight:
161.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

135-137 °C (lit.)

functional group

thioether

SMILES string

CCSc1nnc(N)s1

InChI

1S/C4H7N3S2/c1-2-8-4-7-6-3(5)9-4/h2H2,1H3,(H2,5,6)

InChI key

VWRHSNKTSSIMGE-UHFFFAOYSA-N

General description

2-Amino-5-(ethylthio)-1,3,4-thiadiazole is an inhibitor of copper corrosion. Effect of 2-amino-5-ethylthio-1,3,4-thiadiazole on the copper corrosion in aerated HCl solution has been investigated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effectiveness of corrosion inhibitor films for the conservation of bronzes and gilded bronzes.
Balbo A, et al.
Corrosion Science, 59, 204-212 (2012)
Effects of 2-amino-5-ethylthio-1, 3, 4-thiadiazole on copper corrosion as a corrosion inhibitor in aerated acidic pickling solutions.
Sherif EM and Park S-M.
Electrochimica Acta, 51(28), 6556-6562 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service