Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

334510

Sigma-Aldrich

Potassium hexacyanoplatinate(IV)

Synonym(s):

Dipotassium hexakis(cyano)platinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Pt(CN)6
CAS Number:
Molecular Weight:
429.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

Quality Level

form

crystalline

SMILES string

[K+].[K+].N#C[Pt--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.2K.Pt/c6*1-2;;;/q;;;;;;2*+1;-2

InChI key

QHUVCDBBKULIIT-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service