Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

334464

Sigma-Aldrich

Ammonium hexachlororuthenate(IV)

Synonym(s):

Ruthenium(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2RuCl6
CAS Number:
Molecular Weight:
349.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

powder

SMILES string

[NH4+].[NH4+].Cl[Ru--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2H3N.Ru/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

DHTSBQSPUBOKKI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael W Cooke et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(26), 14703-14711 (2020-06-17)
The undeclared release and subsequent detection of ruthenium-106 (106Ru) across Europe from late September to early October of 2017 prompted an international effort to ascertain the circumstances of the event. While dispersion modeling, corroborated by ground deposition measurements, has narrowed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service