Skip to Content
MilliporeSigma
All Photos(1)

Documents

33420

Sigma-Aldrich

5,6-Diaminouracil sulfate salt

purum, ≥95.0% (HPLC)

Synonym(s):

5,6-Diamino-2,4-dihydroxypyrimidine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O2 · 0.5H2SO4
CAS Number:
Molecular Weight:
191.16
Beilstein:
3781587
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥95.0% (HPLC)

ign. residue

≤1%

mp

≥300 °C

SMILES string

OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service