Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

333611

Sigma-Aldrich

6-Phenyl-1-hexanol

97%

Synonym(s):

Benzenehexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)6OH
CAS Number:
Molecular Weight:
178.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.511 (lit.)

bp

154-155 °C/11 mmHg (lit.)

density

0.953 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCc1ccccc1

InChI

1S/C12H18O/c13-11-7-2-1-4-8-12-9-5-3-6-10-12/h3,5-6,9-10,13H,1-2,4,7-8,11H2

InChI key

FDXBUMXUJRZANT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Torrens
Molecular diversity, 8(4), 365-370 (2004-12-23)
Valence topological charge-transfer (CT) indices are applied to the calculation ofdipole moments. The dipole moments calculated by algebraic and vector semisums of the CT indices are defined. The combination of the CT indices allows the estimation of the dipole moments.
Masahiko Imai et al.
Bioorganic & medicinal chemistry, 23(13), 3788-3795 (2015-04-26)
Obesity is a risk factor associated with several lifestyle-related diseases, for example, diabetes, high blood pressure, hyperlipidemia and cancer. Caffeic acid 2-phenylethyl ester (CAPE, 1), a naturally-occurring compound found in various plants and propolis, which exhibits anti-inflammatory, immunomodulatory and cytotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service