Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

331791

Sigma-Aldrich

Dimethyl exo-tricyclo[4.2.1.02,5]nona-3,7-diene-3,4-dicarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O4
CAS Number:
Molecular Weight:
234.25
MDL number:
UNSPSC Code:
12352103

Assay

99%

SMILES string

COC(=O)C1=C([C@H]2[C@H]3C[C@H](C=C3)[C@@H]12)C(=O)OC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service