Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330892

Sigma-Aldrich

(S)-(−)-Atenolol

99%

Synonym(s):

(−)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH2CH(OH)CH2OC6H4CH2CONH2
CAS Number:
Molecular Weight:
266.34
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

99%

form

solid

optical activity

[α]25/D −16°, c = 1 in 1 M HCl

mp

148-152 °C (lit.)

SMILES string

CC(C)NC[C@H](O)COc1ccc(CC(N)=O)cc1

InChI

1S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17H,7-9H2,1-2H3,(H2,15,18)/t12-/m0/s1

InChI key

METKIMKYRPQLGS-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin F Morris et al.
Chemical physics, 457, 133-146 (2015-08-11)
Molecular dynamics simulations and NMR spectroscopy were used to compare the binding of two β-blocker drugs to the chiral molecular micelle poly-(sodium undecyl-(L)-leucine-valine). The molecular micelle is used as a chiral selector in capillary electrophoresis. This study is part of
Ulisse Garbin et al.
Mediators of inflammation, 2008, 367590-367590 (2008-04-26)
The endothelium plays a key role in the development of atherogenesis and its inflammatory and proliferative status influences the progression of atherosclerosis. The aim of this study is to compare the effects of two beta blockers such as nebivolol and
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service