Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330612

Sigma-Aldrich

3-Methyl-1-cyclohexanecarboxylic acid, mixture of cis and trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10CO2H
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.459 (lit.)

density

0.989 g/mL at 25 °C (lit.)

SMILES string

CC1CCCC(C1)C(O)=O

InChI

1S/C8H14O2/c1-6-3-2-4-7(5-6)8(9)10/h6-7H,2-5H2,1H3,(H,9,10)

InChI key

CIFSKZDQGRCLBN-UHFFFAOYSA-N

Application

3-Methyl-1-cyclohexanecarboxylic acid was employed as a model naphthenic acid for quantitative determination of naphthenic acids in water by liquid chromatography-accurate mass time-of-flight mass spectrometry. It was also used in the synthesis of substituted cyclohexyl carbonyl chlorides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of mefway analogs as ligands for serotonin 5HT1A receptors.
Thio JP, et al.
Medicinal Chemistry Research, 1-7 null
Ralph Hindle et al.
Journal of chromatography. A, 1286, 166-174 (2013-03-23)
This study details the development of a routine method for quantitative analysis of oil sands naphthenic acids, which are a complex class of compounds found naturally and as contaminants in oil sands process waters from Alberta's Athabasca region. Expanding beyond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service