Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330582

Sigma-Aldrich

2-[4-(Dimethylamino)styryl]-1-methylquinolinium iodide

Dye content 97 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21IN2
CAS Number:
Molecular Weight:
416.30
Beilstein:
6102208
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 97%

mp

261 °C (dec.) (lit.)

λmax

524 nm

SMILES string

[I-].CN(C)c1ccc(cc1)\C=C\c2ccc3ccccc3[n+]2C

InChI

1S/C20H21N2.HI/c1-21(2)18-12-8-16(9-13-18)10-14-19-15-11-17-6-4-5-7-20(17)22(19)3;/h4-15H,1-3H3;1H/q+1;/p-1

InChI key

VGLNXCCOYCGRLG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cosimo G Fortuna et al.
European journal of medicinal chemistry, 47(1), 221-227 (2011-11-29)
Almond and VolSurf + modelling procedures allowed the structural design of new di- and mono-heteroaryl-ethylenes. The structural modifications suggested by the molecular modelling were verified by the synthesis of the designed molecules and by the evaluation of their in vitro activities against two
Kullapa Chanawanno et al.
European journal of medicinal chemistry, 45(9), 4199-4208 (2010-07-14)
The series of pyridinium (1-10) and quinolinium (11-20) stilbene benzenesulfonates have been synthesized and their structures were investigated by UV-vis, FT-IR and (1)H NMR spectroscopy. In addition, compound 5 was also determined by single crystal X-ray diffraction technique. The antibacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service