Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330531

Sigma-Aldrich

Sodium 1-naphthalenesulfonate

technical grade, 80%

Synonym(s):

1-Naphthalenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NaO3S
CAS Number:
Molecular Weight:
230.22
Beilstein:
3639038
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

form

powder

SMILES string

[Na+].[O-]S(=O)(=O)c1cccc2ccccc12

InChI

1S/C10H8O3S.Na/c11-14(12,13)10-7-3-5-8-4-1-2-6-9(8)10;/h1-7H,(H,11,12,13);/q;+1/p-1

InChI key

HIEHAIZHJZLEPQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 1-naphthalenesulfonate was employed as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It was also employed as a stationary liquid phase for resolution of dichlorophenol isomers by GC.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-liquid chromatographic resolution of free dichlorophenol isomers on new stationary liquid phases.
Ono, A., and Y. Masuda.
Chromatographia, 17(12), 691-692 (1983)
Ion-Pair High-Performance Liquid Chromatographic Retention Behavior of Copper (II)-1-Oxa-4, 7, 10, 13-tetraazacyclopentadecane Complex.
Chung Y, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 62(3), 336-343 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service