Skip to Content
MilliporeSigma
All Photos(1)

Documents

330116

Sigma-Aldrich

Palladium on carbon

extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E101 NO/W

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:

form

powder

contains

~50% water

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

5 wt. % loading (dry basis)

weight

Unit wt. excludes wt. of H2O

matrix

activated carbon, wet support

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Stille reaction
  • Hydrogenation reactions
  • Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions
  • Deoxygenation
  • Oxidation reactions
  • Coupling reactions
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service