Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329487

Sigma-Aldrich

(−)-8-Phenylmenthol

98%

Synonym(s):

(−)-Phenmenthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O
CAS Number:
Molecular Weight:
232.36
Beilstein:
3651167
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −26°, c = 2 in ethanol

refractive index

n20/D 1.53 (lit.)

density

0.999 g/mL at 20 °C (lit.)

SMILES string

C[C@@H]1CC[C@H]([C@H](O)C1)C(C)(C)c2ccccc2

InChI

1S/C16H24O/c1-12-9-10-14(15(17)11-12)16(2,3)13-7-5-4-6-8-13/h4-8,12,14-15,17H,9-11H2,1-3H3/t12-,14-,15-/m1/s1

InChI key

WTQIZFCJMGWUGZ-BPLDGKMQSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service