Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329290

Sigma-Aldrich

N,N-Dimethyl-2-chloroacetoacetamide

96%

Synonym(s):

2-Chloro-N,N-dimethylacetoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCHClCON(CH3)2
CAS Number:
Molecular Weight:
163.60
Beilstein:
1761325
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.484 (lit.)
n/D (lit.)

bp

95 °C/0.7 mmHg (lit.)

density

1.203 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(=O)C(Cl)C(C)=O

InChI

1S/C6H10ClNO2/c1-4(9)5(7)6(10)8(2)3/h5H,1-3H3

InChI key

QJACRCAPQIYMIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyl-2-chloroacetoacetamide is an active component of Aframomum danielli spice powder.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preservation of Some Physico-Chemical Properties of Soymilk-Based Juice with Aframomum Danielli Spice Powder.
Dauda AO and Adegoke GO.
American Journal of Food Science and Technology, 2(4), 116-121 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service