Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329258

Sigma-Aldrich

6,7-Dimethoxy-2,4-quinazolinedione

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O4
CAS Number:
Molecular Weight:
222.20
Beilstein:
204597
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

>300 °C (lit.)

SMILES string

COc1cc2NC(=O)NC(=O)c2cc1OC

InChI

1S/C10H10N2O4/c1-15-7-3-5-6(4-8(7)16-2)11-10(14)12-9(5)13/h3-4H,1-2H3,(H2,11,12,13,14)

InChI key

KWNQIIMVPSMYEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6,7-Dimethoxy-2,4-quinazolinedione may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service