Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328537

Sigma-Aldrich

4-[(S,S)-2,3-Epoxyhexyloxy]phenyl 4-(decyloxy)benzoate

98%

Synonym(s):

WP 46

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H40O5
CAS Number:
Molecular Weight:
468.62
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal

optical activity

[α]20/D −12°, c = 1 in chloroform-d

SMILES string

CCCCCCCCCCOc1ccc(cc1)C(=O)Oc2ccc(OC[C@@H]3O[C@H]3CCC)cc2

InChI

1S/C29H40O5/c1-3-5-6-7-8-9-10-11-21-31-24-15-13-23(14-16-24)29(30)33-26-19-17-25(18-20-26)32-22-28-27(34-28)12-4-2/h13-20,27-28H,3-12,21-22H2,1-2H3/t27-,28-/m0/s1

InChI key

SDWUPBPBMWMRLN-NSOVKSMOSA-N

General description

4-[(S,S)-2,3-Epoxyhexyloxy]phenyl 4-(decyloxy)benzoate is a ferroelectric liquid crystal. Effect of polymer networks on its smectic-C* phase was studied by high resolution X ray diffraction. Investigations of 4-[(S,S)-2,3-Epoxyhexyloxy]phenyl 4-(decyloxy)benzoate by a polarizing microscope, optical dispersion and UV-VIS spectroscopy have been conducted.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Upindranath Singh
Physical review. E, Statistical, nonlinear, and soft matter physics, 83(6 Pt 1), 061707-061707 (2011-07-30)
High-resolution x-ray diffraction was used to investigate how polymer networks affect the tilt angle in the smectic-C* (SmC*) phase of the ferroelectric liquid crystal 4-[(S,S)-2,3-epoxyhexyloxy]-phenyl-4-(decyloxy)-benzoate (W46 or EPHDBPE). Blends comprising W46, a monomer, and a photoinitiator were prepared and photopolymerized
Investigations of tbe structure of a cholesteric phase with a temperature induced helix inversion and of the succeeding Sc* phase in thin liquid crystal cells
Dierking I, et al.
Liq. Cryst., 13(1), 45-55 null

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service