Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328421

Sigma-Aldrich

1-Pentene

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
1731629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.4 (vs air)

vapor pressure

10.27 psi ( 20 °C)

Assay

90%

autoignition temp.

522 °F

expl. lim.

8.7 %

refractive index

n20/D 1.371 (lit.)

bp

29.9-30.1 °C (lit.)

density

0.641 g/mL at 25 °C (lit.)

SMILES string

CCCC=C

InChI

1S/C5H10/c1-3-5-4-2/h3H,1,4-5H2,2H3

InChI key

YWAKXRMUMFPDSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Merrigan et al.
Organic letters, 1(2), 327-329 (2000-05-24)
[formula: see text] The 13C and 2H kinetic isotope effects for the bromination of 1-pentene with Br2 in CCl4 were determined and interpreted with the aid of calculationally predicted isotope effects. The isotope effects observed are consistent with rate-limiting bromonium
Yu Shan Susanna Wan et al.
Chemical communications (Cambridge, England), (8)(8), 878-879 (2002-07-19)
A zeolite-based microengineered reactor was fabricated and tested for 1-pentene epoxidation over titanium silicalite-1 (TS-1) catalyst, which has been selectively incorporated within the microreactor channel using a new synthesis procedure.
Chen Zhao et al.
Urology, 82(3), 744-744 (2013-07-31)
To analyze the gas generated from the transurethral resection of the prostate (TURP) and transurethral resection of bladder (TURB) tumor. Thirty-six smoke samples were collected from a continuous irrigation suction system during the TURP and the TURB. Then, they were
Takuya Kanemitsu et al.
Organic letters, 5(24), 4541-4544 (2003-11-25)
[reaction: see text] The octenediol linker used during automated oligosaccharide assembly is cleaved by olefin cross-metathesis. Until now, this linker could not be applied to sugars containing azides. A detailed study of the cleavage reaction served as basis for the
Esther Marco et al.
Journal of chromatography. A, 1410, 51-59 (2015-08-06)
A method for the rapid analysis of volatile organic compounds (VOCs) in smoke from tobacco and electronic cigarettes and in exhaled breath of users of these smoking systems has been developed. Both disposable and rechargeable e-cigarettes were considered. Smoke or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service