Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328340

Sigma-Aldrich

2-Methyl-2-propenyl phenyl ether

90%, technical grade

Synonym(s):

Methallyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CH2OC6H5
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

vapor density

5.1 (vs air)

Assay

90%

refractive index

n20/D 1.514 (lit.)

bp

175-176 °C (lit.)

density

0.964 g/mL at 25 °C (lit.)

SMILES string

CC(=C)COc1ccccc1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service