Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328332

Sigma-Aldrich

Dimethylphosphinic chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2P(O)Cl
CAS Number:
Molecular Weight:
112.50
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

204 °C (lit.)

mp

65-71 °C (lit.)

functional group

phosphine oxide

SMILES string

CP(C)(Cl)=O

InChI

1S/C2H6ClOP/c1-5(2,3)4/h1-2H3

InChI key

CVNMBKFJYRAHPO-UHFFFAOYSA-N

General description

Kinetics of pyridinolyses of dimethylphosphinic chloride in acetonitrile was studied. Kinetics and mechanism of the reaction of dimethyl phosphinic chloride with anilines in acetonitrile was studied.

Application

Dimethylphosphinic chloride was used in preparation of dimethyl-ethynylphosphine oxide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dialkylethynylphosphine oxides; synthesis and reactions.
Hagens W, et al.
Rec. Trav. Chim., 88(1), 71-85 (1969)
Kinetics and mechanism of the aminolysis of dimethyl and methyl phenyl phosphinic chlorides with anilines.
Dey NK, et al.
Journal of Physical Organic Chemistry, 22(5), 425-430 (2009)
Pyridinolysis of Diethyl Phosphinic Chloride in Acetonitrile.
Dey NK, et al.
Bull. Korean Chem. Soc., 32(2), 709-712 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service