Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

327859

Sigma-Aldrich

2′,3′-Dideoxy-5-iodouridine

97%

Synonym(s):

5-Iodo-2′,3′-dideoxyuridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2O4
CAS Number:
Molecular Weight:
338.10
Beilstein:
5956836
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

176 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

OC[C@@H]1CC[C@@H](O1)N2C=C(I)C(=O)NC2=O

InChI

1S/C9H11IN2O4/c10-6-3-12(9(15)11-8(6)14)7-2-1-5(4-13)16-7/h3,5,7,13H,1-2,4H2,(H,11,14,15)/t5-,7+/m0/s1

InChI key

OBGFSDPYSQAECJ-CAHLUQPWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′,3′-Dideoxy-5-iodouridine is commonly used as research tool for antiviral and anticancer studies.[1]

Application

2′,3′-Dideoxy-5-iodouridine (5-Iodo-2′,3′-dideoxyuridine) was used in preparation of fluorescence-tagged chain-terminating reagents, T-526, for rapid DNA sequencing.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Prober et al.
Science (New York, N.Y.), 238(4825), 336-341 (1987-10-16)
A DNA sequencing system based on the use of a novel set of four chain-terminating dideoxynucleotides, each carrying a different chemically tuned succinylfluorescein dye distinguished by its fluorescent emission is described. Avian myeloblastosis virus reverse transcriptase is used in a
null
Aldrichimica Acta, 20, 52-52 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service