Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

326186

Sigma-Aldrich

Cyclohexanone cyanohydrin

98%

Synonym(s):

1-Hydroxy-1-cyclohexanecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H10CN
CAS Number:
Molecular Weight:
125.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

132 °C/19 mmHg (lit.)

mp

32-35 °C (lit.)

storage temp.

2-8°C

SMILES string

OC1(CCCCC1)C#N

InChI

1S/C7H11NO/c8-6-7(9)4-2-1-3-5-7/h9H,1-5H2

InChI key

ZDBRPNZOTCHLSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanone cyanohydrin is an aromatic cyanohydrin.

Application

Cyclohexanone cyanohydrin was employed as substrate during high-throughput screening assay for hydroxynitrile lyase activity. It was used in preparation of 1-aminomethyl cyclohexanol via catalytic hydrogenation reaction.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Reduction of Cyclohexanone Cyanohydrin by Lithium Aluminum Hydride.
Nace HR and Smith BB.
Journal of the American Chemical Society, 74(7), 1861-1862 (1952)
Jennifer Andexer et al.
Chemical communications (Cambridge, England), (40)(40), 4201-4203 (2006-10-13)
A high-throughput screening assay for hydroxynitrile lyase activity accepting a wide range of HNL-substrates is presented, which is useful either for enzyme fingerprinting or screening of huge variant libraries generated in metagenome or directed evolution approaches.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service