Skip to Content
MilliporeSigma
All Photos(1)

Documents

326178

Sigma-Aldrich

1-Chlorocarbonyl-1-methylethyl acetate

95%

Synonym(s):

α-Acetoxyisobutyryl chloride, 2-Acetoxy-2-methylpropionyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OCOCH3)COCl
CAS Number:
Molecular Weight:
164.59
Beilstein:
507772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

55-56 °C/6 mmHg (lit.)

density

1.136 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OC(C)(C)C(Cl)=O

InChI

1S/C6H9ClO3/c1-4(8)10-6(2,3)5(7)9/h1-3H3

InChI key

RBTCRFLJLUNCLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 1-chlorocarbonyl-1-methylethyl acetate with 1-aryl ethylene glycols to yield trans chlorohydrin acetates was reported.

Application

1-Chlorocarbonyl-1-methylethyl acetate was used in preparation of α,γ-dichloro alditols.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahesh K Lakshman et al.
Journal of the American Chemical Society, 129(1), 68-76 (2007-01-04)
A diastereoselective synthesis of the nucleoside adducts corresponding to a cis ring-opening of the carcinogen (+/-)-7 beta, 8 alpha-dihydroxy-9 alpha,10 alpha-epoxy-7,8,9,10-tetrahydrobenzo[a]pyrene (BaP DE-2) by 2'-deoxyadenosine and 2'-deoxyguanosine is described. The key intermediate (+/-)-10alpha-amino-7beta,8alpha,9alpha-trihydroxy-7,8,9,10-tetrahydrobenzo[a]pyrene was synthesized by a highly diastereoselective dihydroxylation
Efficient synthesis of some dichloroalditols: direct regioselective chlorination of some unprotected alditols by 1-chlorocarbonyl-1-methylethyl acetate.
Benazza M, et al.
Journal of Carbohydrate Chemistry, 13(7), 967-979 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service