Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

326070

Sigma-Aldrich

Yttrium(III) sulfate octahydrate

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y2(SO4)3 · 8H2O
CAS Number:
Molecular Weight:
610.12
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9% trace metals basis

form

powder

density

2.5 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.[Y+3].[Y+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/3H2O4S.8H2O.2Y/c3*1-5(2,3)4;;;;;;;;;;/h3*(H2,1,2,3,4);8*1H2;;/q;;;;;;;;;;;2*+3/p-6

InChI key

OMBXBIHFXZAMLA-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross K Taggart et al.
Environmental science. Processes & impacts, 20(10), 1390-1403 (2018-09-29)
Coal combustion ash is a promising alternative source of rare earth elements (REE; herein defined as the 14 stable lanthanides, yttrium, and scandium). Efforts to extract REE from coal ash will depend heavily on the location and speciation of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service