Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32596

Sigma-Aldrich

Diallyl pyrocarbonate

≥95.0% (T)

Synonym(s):

(Alloc)2O, Diallyl dicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CHCH2OCO)2O
CAS Number:
Molecular Weight:
186.16
Beilstein:
4390381
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

≥95.0% (T)

refractive index

n20/D 1.434 (lit.)
n20/D 1.434

bp

60 °C/0.05 mmHg (lit.)

density

1.121 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C=CCOC(=O)OC(=O)OCC=C

InChI

1S/C8H10O5/c1-3-5-11-7(9)13-8(10)12-6-4-2/h3-4H,1-2,5-6H2

InChI key

XVSSGIXTKVRGAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diallyl pyrocarbonate or (Alloc)2O is used as a reagent for the introduction of allyloxycarbonyl (Alloc or Aloc) protecting group to primary and secondary amines. In peptide synthesis, it is also used to protect amino acid functional groups due to its stability to the acidic and basic conditions. Various amino functional groups such as amino sugars and nucleosides are also protected by using diallyldicarbonate reagent.

Other Notes

Reagent for the convenient introduction of the N-allyloxycarbonyl (N-Alloc) amino protection ; Allyl carbamate protections can be readily cleaved by palladium(0) reagents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diallyl dicarbonate. A convenient reagent for the synthesis of allyl carbamates.
Barcelo G, et al.
Tetrahedron Letters, 28(47), 5809-5810 (1987)
O. Dangles et al.
The Journal of Organic Chemistry, 52, 4984-4984 (1987)
G. Sennyey et al.
Tetrahedron Letters, 28, 5809-5809 (1987)
H. Kunz et al.
Angewandte Chemie (International Edition in English), 96, 426-426 (1984)
N-[(Allyloxycarbonyl) oxy] maleimide-styrene Copolymer
N Carmen, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-2 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service