Skip to Content
MilliporeSigma
All Photos(1)

Documents

324604

Sigma-Aldrich

Chloromethyl phenyl sulfone

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2Cl
CAS Number:
Molecular Weight:
190.65
Beilstein:
2046644
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

mp

51-53 °C (lit.)

SMILES string

ClCS(=O)(=O)c1ccccc1

InChI

1S/C7H7ClO2S/c8-6-11(9,10)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

NXAIQSVCXQZNRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service