Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

322482

Sigma-Aldrich

4-Hydroxy-2,5-dimethyl-3(2H)-furanone

95%

Synonym(s):

2,5-Dimethyl-4-hydroxy-3(2H)-furanone, Furaneol, Strawberry furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
Beilstein:
1281357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

73-77 °C (lit.)

SMILES string

CC1OC(C)=C(O)C1=O

InChI

1S/C6H8O3/c1-3-5(7)6(8)4(2)9-3/h3,8H,1-2H3

InChI key

INAXVXBDKKUCGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Hauck et al.
Journal of agricultural and food chemistry, 51(16), 4753-4756 (2004-01-07)
The formation of 4-hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF) by Zygosaccharomyces rouxii was studied in yeast-peptone-dextrose medium containing d-fructose 1,6-diphosphate under various culture conditions. Cell growth and HDMF production was heavily dependent on medium pH and sodium chloride concentration. Higher pH values of the
Yu Wang et al.
Journal of agricultural and food chemistry, 56(16), 7405-7409 (2008-07-03)
The caramel-like aroma compound, 2,5-dimethyl-4-hydroxy-3(2H)-furanone (DMHF) was quantified and verified by HPLC and GC-MS in the Maillard reaction based on methylglyoxal (MG). The reaction was performed in the 0.5 M phosphate buffer by heating MG with or without either glycine
Nonjabulo P Gule et al.
Water research, 47(3), 1049-1059 (2012-12-25)
The 3(2H) furanone derivative 2,5-dimethyl-4-hydroxy-3(2H)-furanone (DMHF) was investigated for its antimicrobial and cell-adhesion inhibition properties against Klebsiella pneumoniae Xen 39, Staphylococcus aureus Xen 36, Escherichia coli Xen 14, Pseudomonas aeruginosa Xen 5 and Salmonella typhimurium Xen 26. Nanofibers electrospun from
Stefan Lunkenbein et al.
Journal of experimental botany, 57(10), 2445-2453 (2006-06-27)
A complex mixture of hundreds of substances determines strawberry (Fragaria x ananassa) aroma, but only approximately 15 volatiles are considered as key flavour compounds. Of these, 4-hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF) is regarded as the most important, but it is methylated further by
M C Rodríguez Dodero et al.
Journal of agricultural and food chemistry, 58(2), 990-997 (2009-12-22)
This article shows the results obtained in the study of the extraction profiles from oak wood to distillate of several compounds, low molecular weight phenolics, and furanic derivatives, and the relationship of their contents with those found in commercial sherry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service