Skip to Content
MilliporeSigma
All Photos(1)

Documents

320161

Sigma-Aldrich

Benzyl chloride

reagent grade, 97%

Synonym(s):

α-Chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
471308
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

reagent grade

vapor density

4.36 (vs air)

vapor pressure

10.3 mmHg ( 60 °C)
7 mmHg ( 55 °C)

Assay

97%

autoignition temp.

1085 °F

contains

0.25% propylene oxide as inhibitor

expl. lim.

14 %

refractive index

n20/D 1.538 (lit.)

bp

177-181 °C (lit.)

mp

−43 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

ClCc1ccccc1

InChI

1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

KCXMKQUNVWSEMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylvie Mavel et al.
Bioorganic & medicinal chemistry, 20(4), 1388-1395 (2012-02-04)
The dopamine transporter (DAT) plays a pivotal role in the regulation of dopamine neurotransmission, and is involved in a number of physiological functions and brain disorders. Furthermore the DAT analysis by molecular imaging techniques is a useful tool for the
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service