Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

319716

Sigma-Aldrich

(1R)-(+)-Camphorsulfonylimine

99%

Synonym(s):

(3aR)-(+)-4,5,6,7-Tetrahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole-2,2-dioxide, (7R)-(+)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.01,5]dec-4-ene 3,3-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2S
CAS Number:
Molecular Weight:
213.30
Beilstein:
85297
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D +34°, c = 1 in chloroform

mp

229-230 °C (lit.)

SMILES string

CC1(C)C2CC[C@@]13CS(=O)(=O)N=C3C2

InChI

1S/C10H15NO2S/c1-9(2)7-3-4-10(9)6-14(12,13)11-8(10)5-7/h7H,3-6H2,1-2H3/t7?,10-/m0/s1

InChI key

ZAHOEBNYVSWBBW-MHPPCMCBSA-N

Application

(R) and (S) forms are precursors to chiral (camphorsulfonyl)oxaziridines employed in the asymmetric hydroxylation of ester and amide enolates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 51, 2402-2402 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service