Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

318523

Sigma-Aldrich

(S)-(+)-γ-Ethoxycarbonyl-γ-butyrolactone

93%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O4
CAS Number:
Molecular Weight:
158.15
MDL number:
UNSPSC Code:
12352103

Assay

93%

optical activity

[α]25/D +7.5°, neat

refractive index

n20/D 1.447 (lit.)

bp

135-140 °C/10 mmHg (lit.)

density

1.163 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)[C@@H]1CCC(=O)O1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service