Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

318272

Sigma-Aldrich

Xenon difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
XeF2
CAS Number:
Molecular Weight:
169.29
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

3.8 mmHg ( 25 °C)

mp

129 °C (lit.)

density

4.32 g/mL at 25 °C (lit.)

SMILES string

F[Xe]F

InChI

1S/F2Xe/c1-3-2

InChI key

IGELFKKMDLGCJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Very useful fluorination agent.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorination with XeF(2).(1) 44. Effect of Geometry and Heteroatom on the Regioselectivity of Fluorine Introduction into an Aromatic Ring.
Marko Zupan et al.
The Journal of organic chemistry, 63(3), 878-880 (2001-10-24)
Ariela W Kaspi et al.
Inorganic chemistry, 47(1), 5-7 (2007-12-07)
Palladium(II) aryliodo complexes bearing chelating diphosphine ligands react with XeF2, giving iodoarene and rare palladium(II) difluoro complexes. The reaction is general with regard to the aryl group, with even C6F5-I undergoing facile reductive elimination from a Pd center.
Tsung Yi Chiang et al.
Journal of synchrotron radiation, 17(1), 69-74 (2009-12-24)
The synchrotron radiation (SR) stimulated etching of silicon elastomer polydimethylsiloxane (PDMS) using XeF(2) as an etching gas has been demonstrated. An etching system with differential pumps and two parabolic focusing mirrors was constructed to perform the etching. The PDMS was
Minseob Kim et al.
Nature chemistry, 2(9), 784-788 (2010-08-24)
The application of pressure, internal or external, transforms molecular solids into extended solids with more itinerant electrons to soften repulsive interatomic interactions in a tight space. Examples include insulator-to-metal transitions in O(2), Xe and I(2), as well as molecular-to-non-molecular transitions
Neil Vasdev et al.
Inorganic chemistry, 49(19), 8997-9004 (2010-08-31)
The existence of the trifluoroxenate(II) anion, XeF(3)(-), had been postulated in a prior NMR study of the exchange between fluoride ion and XeF(2) in CH(3)CN solution. The enthalpy of activation for this exchange, ΔH(⧧), has now been determined by use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service