Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

318213

Sigma-Aldrich

Sorbitan monolaurate

Synonym(s):

Span® 20

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O6
CAS Number:
Molecular Weight:
346.46
Beilstein:
8355657
EC Number:
MDL number:
UNSPSC Code:
12352300

refractive index

n20/D 1.474 (lit.)

density

1.032 g/mL at 25 °C (lit.)

HLB

8

SMILES string

CCCCCCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O

InChI

1S/C18H34O6/c1-2-3-4-5-6-7-8-9-10-11-16(21)23-13-15(20)18-17(22)14(19)12-24-18/h14-15,17-20,22H,2-13H2,1H3/t14-,15?,17+,18+/m0/s1

InChI key

LWZFANDGMFTDAV-WYDSMHRWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Span is a registered trademark of Croda International PLC

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service