Skip to Content
MilliporeSigma
All Photos(1)

Documents

317721

Sigma-Aldrich

Dicyclohexylmethanol

98%

Synonym(s):

Dicyclohexylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2CHOH
CAS Number:
Molecular Weight:
196.33
Beilstein:
1926669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

bp

154 °C/12 mmHg (lit.)

mp

58-64 °C (lit.)

SMILES string

OC(C1CCCCC1)C2CCCCC2

InChI

1S/C13H24O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-14H,1-10H2

InChI key

DEFUSPNGFCCTEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Araceli Rivera-Pérez et al.
Food chemistry, 321, 126727-126727 (2020-04-14)
Alkenylbenzenes are natural toxins with genotoxic and carcinogenic effects in rodents, which are highly present in condiments frequently consumed. The aim of this study was the development of the first multi-analyte method for the determination of eight alkenylbenzenes (eugenol, methyl
Martin Eckardt et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(7), 1421-1437 (2018-03-15)
Polyphenylsulfone (PPSU) is a new material for the production of baby bottles. PPSU is a polyether plastic formally composed of bisphenol S (BPS) and 4,4'-dihydroxybiphenyl (DHBP), which both have slight endocrine activities in in vitro tests. So far, little is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service