Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

311219

Sigma-Aldrich

Dibenzyl dicarbonate

92%

Synonym(s):

Dibenzyl pyrocarbonate, Z2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CO2CH2C6H5)2O
CAS Number:
Molecular Weight:
286.28
Beilstein:
4324796
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

92%

form

solid

mp

29-32 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

functional group

carbonate
phenyl

storage temp.

−20°C

SMILES string

O=C(OCc1ccccc1)OC(=O)OCc2ccccc2

InChI

1S/C16H14O5/c17-15(19-11-13-7-3-1-4-8-13)21-16(18)20-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

FHRRJZZGSJXPRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the introduction of the Z-protecting group in amino acids and other amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 44, 5563-5563 (1988)
Journal of the Chemical Society. Perkin Transactions 1, 1905-1905 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service