Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

309915

Sigma-Aldrich

3,7-Dimethyl-3-octanol

98%

Synonym(s):

Tetrahydrolinalool

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3C(CH3)(OH)C2H5
CAS Number:
Molecular Weight:
158.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Assay

98%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

71-73 °C/6 mmHg (lit.)

density

0.826 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCC(C)(O)CCCC(C)C

InChI

1S/C10H22O/c1-5-10(4,11)8-6-7-9(2)3/h9,11H,5-8H2,1-4H3

InChI key

DLHQZZUEERVIGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The chlorooxidation of 3,7-dimethyl-3-octanol (tetrahydrolinalool) was studied.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark B Erman et al.
Chemistry & biodiversity, 5(6), 910-919 (2008-07-12)
Alpha-pinene and beta-pinene are abundantly represented in nature; they are obtained from renewable sources and are irreplaceable synthons for commercial-scale production of other terpenes. In a well-known process practiced by Millennium Specialty Chemicals (MSC), hydrogenation of pinenes gives cis- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service