Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

309885

Sigma-Aldrich

2,4-Dichloro-1-fluorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3F
CAS Number:
Molecular Weight:
164.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.525 (lit.)

bp

172-174 °C (lit.)

density

1.409 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Cl)cc1Cl

InChI

1S/C6H3Cl2F/c7-4-1-2-6(9)5(8)3-4/h1-3H

InChI key

BDJZCCWUSOZUQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service