Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

309680

Sigma-Aldrich

5,6-Dihydro-4-hydroxy-6-methyl-2H-pyran-2-one

Synonym(s):

(±)-Gerberin aglycone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

126-128 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1CC(O)=CC(=O)O1

InChI

1S/C6H8O3/c1-4-2-5(7)3-6(8)9-4/h3-4,7H,2H2,1H3

InChI key

PXNRZJNTMDCHFR-UHFFFAOYSA-N

Application

5,6-Dihydro-4-hydroxy-6-methyl-2H-pyran-2-one was used in the synthesis of spiro-γ/δ-dilactone as a mixture of diastereomers.
Reactant involved in:
  • Synthesis of spiroheterocycles with fused heterosystems
  • Preparation of insecticidal heterolignans
  • Cyclization of aminoanilino lactones on diazotization

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hang W.
Tetrahedron Letters, 41(15), 2523-2527 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service