Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

309389

Sigma-Aldrich

Nafion NR50

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203
NACRES:
NA.23

form

pellets

eq. wt.

≤1250

capacity

≥0.8 meq/g ion exchange capacity

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

General description

Nafion NR50 is a perfluorosulfonic acid nafion resin that is majorly used as a strong solid acid (H0 -12) with high thermal stability and chemical resistance.

Application

Nafion NR50 can be used for a variety of applications such as:
  • a solid acid-catalyst in the synthesis of α-tocopherol
  • a catalyst for cellulose hydrolysis
  • a reference catalyst in the preparation of carbon-based solid acid catalysts

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of a carbon-based solid acid catalyst by sulfonating activated carbon in a chemical reduction process
Liu X, et al.
Molecules (Basel), 15(10), 7188-7196 (2010)
Catalytic hydrolysis of cellulose to levulinic acid by partly replacing sulfuric acid with Nafion NR50 catalyst
Xu Y, et al.
Chemosphere, 9(3), 609-616 (2019)
Catalytic performance of Nafion/SiO2 nanocomposites for the synthesis of alpha-tocopherol
Wang H and Xu B
Applied Catalysis A: General, 275(1-2), 247-255 (2004)
Min-Ho Park et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4308-4314 (2015-06-24)
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use
Martta Asikainen et al.
Bioresource technology, 191, 300-305 (2015-05-26)
Bio-based fuels are becoming more and more important due to the depleting fossil resources. The production of biodiesel from algae oil is challenging compared to terrestrial vegetable oils, as algae oil consists of polar fatty acids, such as phospholipids and

Articles

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service