Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

307424

Sigma-Aldrich

Silver heptafluorobutyrate

97%

Synonym(s):

Heptafluorobutyric acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2CO2Ag
CAS Number:
Molecular Weight:
320.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

292-294 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)O[Ag]

InChI

1S/C4HF7O2.Ag/c5-2(6,1(12)13)3(7,8)4(9,10)11;/h(H,12,13);/q;+1/p-1

InChI key

GPNIJXACCBKBEP-UHFFFAOYSA-M

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sławomir Wybraniec et al.
Journal of chromatography. A, 1216(41), 6890-6899 (2009-09-08)
Polar betacyanin pigments together with betaxanthins from ripe cactus fruits of Hylocereus polyrhizus (Cactaceae) were fractionated by means of preparative ion-pair high-speed countercurrent chromatography (IP-HSCCC) also using the elution-extrusion (EE) approach for a complete pigment recovery. HSCCC separations were operated
Jan Busch et al.
Environmental pollution (Barking, Essex : 1987), 158(5), 1467-1471 (2010-01-08)
Polyfluoroalkyl compounds (PFCs) are widely used in industry and consumer products. These products could end up finally in landfills where their leachates are a potential source for PFCs into the aqueous environment. In this study, samples of untreated and treated
John L Butenhoff et al.
Reproductive toxicology (Elmsford, N.Y.), 33(4), 513-530 (2011-09-01)
Sequential 28-day and 90-day oral toxicity studies were performed in male and female rats with ammonium perfluorobutyrate (NH(4)(+)PFBA) at doses up to 150 and 30mg/kg-d, respectively. Ammonium perfluorooctanoate was used as a comparator at a dose of 30mg/kg-d in the
Marta Llorca et al.
The Science of the total environment, 431, 139-150 (2012-06-12)
Water has been identified as one of the main routes of human exposure to perfluoroalkyl substances (PFASs). This work assessed the presence of 21 PFASs along the whole water cycle using a new fast and cost effective analytical method based
V Gellrich et al.
Chemosphere, 87(9), 1052-1056 (2012-03-07)
Perfluorinated compounds (PFCs) can be detected worldwide in both, soil and water. In order to study the leaching behavior of this heterogeneous group of compounds in soil, flow-through column experiments have been conducted. Ten perfluoro carboxylates and four perfluoro sulfonates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service