Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

307408

Sigma-Aldrich

Lithium diisopropylamide mono(tetrahydrofuran) solution

1.5 M in cyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22LiNO
CAS Number:
Molecular Weight:
179.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

form

liquid

concentration

1.5 M in cyclohexane

density

0.816 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li+].C1CCOC1.CC(C)[N-]C(C)C

InChI

1S/C6H14N.C4H8O.Li/c1-5(2)7-6(3)4;1-2-4-5-3-1;/h5-6H,1-4H3;1-4H2;/q-1;;+1

InChI key

YTJXGDYAEOTOCG-UHFFFAOYSA-N

Application

Lithium diisopropylamide mono(tetrahydrofuran) can be used:
  • As an initiator in the anionic polymerization of D,L-lactide.
  • In one of the key synthetic steps for the preparation of phenylacetic acid component of muscarinic M3 receptor antagonist.
  • In the synthesis of new sexithiophene derivatives, which are used as electron donor materials in bilayer photovoltaic devices.

Other Notes

may contain a small amount of THF and ethylbenzene

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, hearing organs

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective synthesis of the acid part of a new muscarinic M3 receptor antagonist
Mitsuya M, et al.
Tetrahedron, 56(51), 9901-9907 (2000)
Synthesis and properties of end-capped sexithiophenes incorporating the ethylene dithiothiophene unit
Mason CR, et al.
Journal of Materials Chemistry, 15(14), 1446-1453 (2005)
Anionic polymerization of D, L-lactide initiated by lithium diisopropylamide
Bhaw-Luximon A, et al.
Polymer, 42(24), 9651-9656 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service